Order Entry
Switzerland
ContactUsLinkComponent
 
Quick-RNA™ Kits
 
undefined
Quick-RNA™ Kits

 

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Micro
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Sample size:
    ≤1×10⁶ Cells or 5 mg Tissue
  • Description:
    Quick-RNA™ Microprep Kit
  • Sample capacity:
    10 µg
  • Yield:
    Microprep: 10 µg RNA (binding capacity), ≥6 µl (elution volume)

 

Frequently Bought Together