Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Super Broth

Super broth, 24 months, 1 L, 1.000 mL
Catalog # MSPP340-018-131
Marketsource
Super Broth
Catalog # MSPP340-018-131
Marketsource
CAS Number:  
Super Broth
Super Broth

Super broth, 24 months, 1 L, 1.000 mL

CAS Number:  
Price
43.06 SFr.
Log in for availability
1 * 1,000 ml
Product Details & Documents

Super Broth is used for the mass cultivation of Escherichia coli for high yield protein and plasmid DNA production.

  • Has 220% more peptone and 300% more yeast extract than LB

Super Broth is the best commonly used E.coli growth medium for plasmid production. Super Broth has 220% more peptone and 300% more yeast extract than LB. The major nutrients in peptone and yeast extract are still in excess well after cells reach saturation. Super Broth is used for plasmid or protein production.

Sterile Filtered
UNSPSC: 41106200
Harmonisation Code: 3822.00.5090
ECCN: EAR99
Endotoxin and performance tested

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Caution: Research use only.

Specifications

Specifications

  • Pk:
    1.000 mL
  • Description:
    Super broth
  • Shelf life:
    24 months
  • Volume:
    1 L
  • Cat. No.:
    MSPP340-018-131
  • Sterility:
    Autoclave sterilized
  • Shipping conditions:
    Ambient temperature

Specifications