Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Primary Human Microvascular Endothelial Cells

Primary Human Microvascular Endothelial Cells

Human microvascular endothelial cells, Cryopreserved, Single donor, 1 KIT
Catalog # CELSCSC2M1
Marketsource
Supplier:   CELL SYSTEMS
Primary Human Microvascular Endothelial Cells
Catalog # CELSCSC2M1
Supplier:  CELL SYSTEMS
Marketsource
CAS Number:  
Primary Human Microvascular Endothelial Cells

Human microvascular endothelial cells, Cryopreserved, Single donor, 1 KIT

CAS Number:  
Price
SFr. 454.93
Log in for availability
1 * 1 KIT
Product Details & Documents

Our primary human cells offer unmatched consistency and a more biologically relevant tool than other cell types. Our cells are antibody-free and come from a deep inventory to help researchers with reproducibility and improved human insights.

  • Highly proliferative and healthy (~85% viability)
  • Antibody free and more biologically relevant
  • Offered with high cell count (~1 million cells per vial)
  • Well-cited in scientific journals
  • Ideal for 2D and 3D cultures, microfluidic devices and co-cultures

Primary Human Dermal Microvascular Endothelial Cells (CSC2M1) were isolated from dermis of neonatal donor. This cell is antibody free. Each vial will initiate a 25 to 75 cm² cell culture. These primary cells are isolated, frozen and qualified using Cell Systems Certified Media and Reagents--which are specified for use in the research environment. Cell Systems primary cells are exclusively from normal, site-specific human tissue specimen, and available in frozen vial or proliferating flask.

These cells were originated using Cell Systems Complete Serum-Free Medium (SF-4Z0-500), and subsequently grown and passaged in Cell Systems Complete Medium (4Z0-500). They are available at Passage 3 (<12 cumulative population doublings) cryopreserved in Cell Systems Cell Freezing Medium™ (4Z0-705). This vial will initiate a Passage 4 cell culture in a 75 cm² flask. Each vial contains more than 1×106 cells.

Each vial of cells is shipped with vials of Bac-Off® (antibiotic) and CultureBoost™ (animal derived growth factors) or CultureBoost-R™ (human recombinant growth factors) at no additional cost.

Standard Tests:
HIV Test (donor level HIV AB EIA) - Negative
HIV PCR Test (frozen cell pool by CLIA Licensed Clinical Lab) - Negative
Mycoplasma spp. (frozen cells) (ATCC method by CLIA Licensed Clinical Lab) - Negative.

Characterisation:
Inducible expression of CD36 >95% positive by immunofluorescence
Cytoplasmic VWF >95% positive by immunofluorescence
Cytoplasmic uptake of Di-L-Ac-LDL >95% positive by immunofluorescence.

Delivery information: Shipped on dry ice.

Caution: For research use only.

Specifications

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    1 KIT
  • Single Donor/Pooled:
    Single donor
  • Description:
    Human microvascular endothelial cells
  • Cat. No.:
    CELSCSC2M1
  • Storage conditions:
    Upon receipt, store in liquid nitrogen
  • Storage buffer:
    Frozen in Cell Systems cell freezing medium (4Z0705)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.