Order Entry
Switzerland
ContactUsLinkComponent
Norepinephrine (NE) ELISA Kit
Norepinephrine (NE) ELISA Kit
Catalog # ABCAAB287789-96
Supplier:  Abcam
CAS Number:  
undefined
Norepinephrine (NE) ELISA Kit
Catalog # ABCAAB287789-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Competitive
  • Description:
    Norepinephrine (NE) ELISA Kit, 100 assays

Specifications

About this item

A competitive ELISA kit for quantify Norepinephrine biological fluids.

  • Easy, convenient and time-saving method to measure the level of Norepinephrine in serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 15,625 to 1000 pg/ml
  • Sensitivity: <9,375 pg/ml
  • Assay Precision: Intra-Assay: CV <8%; Inter-Assay: CV <10%
  • This assay has high sensitivity and excellent specificity for detection of Norepinephrine. No significant cross-reactivity or interference between Norepinephrine and analogues was observed

Norepinephrine (NE) is a precursor of epinephrine that is secreted by the adrenal medulla and is a well-known central and autonomic neurotransmitter. Norepinephrine is the principal transmitter of most postganglionic sympathetic fibers and of the diffuse projection system in the brain arising from the locus ceruleus. Norepinephrine is also found in plants and is used pharmacologically as a sympathomimetic. BioVision’s Norepinephrine ELISA kit is a competitive ELISA assay for the quantitative measurement of Norepinephrine in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of Norepinephrine captured from the samples.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.