ContactUsLinkComponent
Solus™ CCS Safety Glass, Scotchgard™ Anti-Fog/Anti-Scratch Coating
Solus™ CCS Safety Glass, Scotchgard™ Anti-Fog/Anti-Scratch Coating
Catalog # MMMASCCS07SGAF-GRN
Supplier:  3M
undefined
Solus™ CCS Safety Glass, Scotchgard™ Anti-Fog/Anti-Scratch Coating
Catalog # MMMASCCS07SGAF-GRN
Supplier:  3M
Specifications
  • Type:
    Scotchgard™ anti-fog/anti-scratch
  • Colour:
    Green
  • Description:
    Solus™ CCS safety glasses
  • Lenses:
    I/O grey, PC
  • Pk:
    1
Specifications
About this item
3M™ Solus™ CCS Safety Glasses are rimless safety glasses that feature lime green temples with a Corded Control System (CCS) for attaching corded earplugs. The polycarbonate lens features a Scotchgard™ anti-fog and anti-scratch coating on both sides for increased durability and improved vision.
  • Modern safety glasses with Cord Control System (CCS) to keep eyewear and corded earplugs attached and ready to use
  • Designed for workers who put on and take off eyewear frequently throughout the day
  • Scotchgard™ anti-fog coating provides superior anti-fog and anti-scratch properties
  • Foam gasket available to help limit eye exposure to debris
  • Suitable for all non-textile 3M™ corded earplugs
  • Frame marking: 3M EN 166 FT CE
  • 3M™ internal testing per EN168 test method when compared with traditional anti-fog coatings
For users who are intermittently exposed to noise, use the Solus™ CCS Safety Glasses. These rimless safety glasses feature lime green temples that have a Corded Control System (CCS), which allows users to keep certain E-A-R™ Corded Earplugs attached and ready for use at all times. The unique temple design holds the cord of the earplugs securely in place and untangled. When the corded earplugs are not in use, the CCS feature allows them to act as a lanyard for carrying protective eyewear around the neck.
The polycarbonate lens features a Scotchgard™ anti-fog and anti-scratch coating for superior anti-fog protection and increased durability that resists fogging for longer than traditional anti-fog coatings, even after up to 25 washings with water.