Order Entry
Switzerland
ContactUsLinkComponent
CMO Stereo Zoom Microscope, SM7
CMO Stereo Zoom Microscope, SM7
Supplier:  MOTIC
New Product
Certificates
undefined
CMO Stereo Zoom Microscope, SM7
Supplier:  MOTIC
New Product

Specifications

  • Catalog No:
  • MOTI1100201800041
  • MOTI1100201800051
  • Working distance:
  • 90 mm
  • 81 mm
  • Focusing system:
  • Coarse with tension adjustment
  • Coarse with tension adjustment
  • Stand:
  • Fixed-arm stand with transmitted illumination
  • Fixed-arm stand with transmitted illumination
  • Electrical:
  • 110 - 240 V
  • 110 - 240 V
  • W×D×H:
  • 367×290×372 mm
  • 367×290×372 mm
  • Weight:
  • 7,4 kg
  • 7,5 kg

Specifications

Frequently Bought Together