PolarPack® Foam Brick, Ice Pack Refrigerant Ice Bricks
SCATFPP24
- Seq:05
- W×D×H:165×114×38 mm
- Pk:768
- Weight:680 g (24 oz.)
PolarPack® gel brick coolant pack refrigerants provide thermal solutions for temperature critical packages during transit and are an alternative to wet ice packs.
- Broad selection available
- Reliable temperature protection
- Never change shape during freezing
Available in a broad selection of styles and a multitude of sizes, they’re designed to provide reliable temperature protection for a vast array of products and packages.
These semi-rigid foam bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.