Order Entry
Switzerland
Orders LinkContactUsLinkComponent
MultiPlate Genie® Microplate Shaker

MultiPlate Genie® Microplate Shaker

MultiPlate Genie, CH-plug
Catalog # 444-1432
MultiPlate Genie® Microplate Shaker
Catalog # 444-1432
MultiPlate Genie® Microplate Shaker
MultiPlate Genie® Microplate Shaker

MultiPlate Genie, CH-plug

Price
SFr. 1’830.00
Log in for availability
1 * 1 ST
Product Details & Documents

Fully programmable microplate shaker for four plates (8 with optional tray).

  • Provides consistent well-to-well and plate-to-plate mixing for 96 and 384-well microplates, deep well plates, or PCR plates
  • Will accept PCR Work up racks with standard microplate footprint for use with PCR tubes and strips
  • Featuring programmable speed, time, pulsing/interval mixing - for accurate and repeatable results

Digital true vortex mixing microplate shaker handles up to 4 microplates, (or up to 8 plates with the optional accessory tray). The combined speed and 1 mm orbit provide consistant well to well and plate to plate uniformity, even in 384-well plates. Featuring programable speed, time and continuous or pulse time, it’s ideal for achieving accurate and repeatable results.

The Pulse feature allows rapid pulsing or timed ''on/off'' mixing for interval mixing. Ideal for use in PCR preparation, digestions, bead mixing, ELISA, EIA, and colorimetric assay applications.

Ordering information: Optional stacking tray accessory for additional 4 microplates (giving total of 8) is available.

Delivery information: Supplied with tray for 4 microplates.

Specifications
  • Description:
    MultiPlate Genie, CH-plug
  • Seq:
    03
  • Cat. No.:
    444-1432
  • Orbit:
    1,0 mm
  • Timer:
    1 - 99 min or continuous
  • Pulse ON/OFF:
    0,1 s - 99 min
  • Speed range:
    2700
  • Model:
    MultiPlate Genie
  • W×D×H:
    250×380×150 mm
  • No. of microplates held:
    4 (or 8 microplates with optional tray)
  • Weight:
    6 kg
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Stacking tray for MultiPlate Genie
    Stacking tray for MultiPlate Genie
    SCIISI-4110
    Log in for availability
    1 * 1 ST - SFr. 281.00