Jenway® Conductivity Cells
Supplier: Cole-Parmer
Jump to:
About this item
Cells for conductivity measurement.
- For use with benchtop and portable meters
Compatible with Jenway meters.
2 Options Available Below
Product Details & Documents
Cells for conductivity measurement.
- For use with benchtop and portable meters
Compatible with Jenway meters.
Documents
Manual Conductivity Cell
Manual
TDS Epoxy Bodied Conductivity Probe
Datasheet
Product Family Options
Product Information
- TypeTemp.MaterialØ×LForAvailabilityPrice
- JENW99959-67General purpose, epoxy-bodied conductivity probe, K = 1, for benchtop meters, 1 m cable length (cell constant K = 1 ± 20%)General purpose, epoxy-bodied conductivity probe, K = 1, for benchtop meters, 1 m cable length (cell constant K = 1 ± 20%)Conductivity0...80 °CEpoxy12×120 mmJenway® meters 4510, 4520 and 3540Log in for availability1 * 1 STSFr. 461.00
- JENW99959-64General purpose, glass-bodied conductivity probe, K = 0,1, for benchtop meters, 1 m cable length (cell constant K = 0,1 ± 20%)General purpose, glass-bodied conductivity probe, K = 0,1, for benchtop meters, 1 m cable length (cell constant K = 0,1 ± 20%)Conductivity0...100 °CGlass12×120 mmJenway® meters 4510, 4520 and 3540Log in for availability1 * 1 STSFr. 416.00