Order Entry
Switzerland
ContactUsLinkComponent
 
Water for cleaning, sterile
Catalog # 911660ZT
Supplier:  VWR Chemicals
CAS Number:  
undefined
Water for cleaning, sterile
Catalog # 911660ZT
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bottle, screw cap
  • Pk:
    11
  • Seq:
    2752
  • Pk Info:
    12x500 ml

Specifications

About this item

 :
 :18.02 g/mol
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332