Exact volume transfer pipettes, Samco™, Thermo Scientific
Supplier: SAMCO
Certificates
About this item
The exact volume transfer pipettes are used for dispensing reagents of small samples.
- Polyethylene
- Non sterile
- Shatterproof
- Ideal for home and POC (Point of Care) testing
- OEM applications
Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage. Useful in clinical laboratories. Custom design options available to meet your specifications.
Specifications
- Catalog No:
- SAMC966NL
- SAMC783NL
- SAMC784NL
- SAMC788NL
- SAMC785NL
- SAMC786NL
- SAMC191NL
- SAMC967NL
- SAMC930NL
- SAMC787NL
- SAMC941NL
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)