Order Entry
Switzerland
ContactUsLinkComponent
Methanol bio-based ≥99.5%, GPR RECTAPUR®
Flammable
Toxic
Health
Methanol bio-based ≥99.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Toxic
Health
Methanol bio-based ≥99.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (GC):
    min. 99.5%
  • Appearance:
    clear colourless liquid
  • IR Spectrum:
    Passes test
  • Acidity (meq/g):
    max. 0.0002
  • Boiling point:
    64 - 65°C
  • Colouration (APHA):
    max. 10
  • Acetone:
    max. 10 ppm
  • Evaporation residue:
    max. 50 ppm
  • Water :
    max. 0.2%

Specification Test Results

Related Information

Frequently Bought Together