Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Media for Microbiology
Prepared Media for Microbiology
Culture media in bottles
Culture media in bottles
Culture media in bottles
Catalog # HIMEFD057-5VL
Supplier:
HIMEDIA
CAS Number:
Expand image
Culture media in bottles
Catalog # HIMEFD057-5VL
Supplier:
HIMEDIA
CAS Number:
Specifications
Pk:
5
Description:
TTC solution 1%
Specifications
About this item
Product Details
Product Details