ContactUsLinkComponent
Interconnect mat cord, SCS
Interconnect mat cord, SCS
Catalog # 115-3047
Supplier:  DESCO
Interconnect mat cord, SCS
Catalog # 115-3047
Supplier:  DESCO
Specifications
  • Colour:
    Black
  • Description:
    Interconnect mat cord, with 10 mm male stud snap fasteners
  • Length:
    10'
  • Pk:
    1
Specifications
About this item
Daisy-chains two worksurface mats for the monitoring of two workstations with the 724 workstation monitor.
  • Reduces inadvertent snagging that may disconnect the cord from the worksurface mat
  • No resistor
This monitor/table mat interconnect cord is a straight cord with male stud snap fasteners at each end, and no resistors.
Certifications: Meets ANSI/ESD S20.20 and Grounding standard ANSI/ESD S6.1.