Surface and mat cleaner, Reztore®
Codice catalogo 115-3048
Nome del fornitore: DESCO
Specifiche
- Description:Surface and mat cleaner, 950 ml
- Packed:Reusable spray bottle
- Pk:1
Specifiche
Informazioni su questo articolo
Reztore® Surface and mat cleaners removes dust, grease, grime, fingerprints, solder flux and other contaminants from ESD mats and other surfaces.
- One quart trigger spray bottle
- New alcohol-free formula
- Non-hazardous, non-flammable green chemistry formula
- No silicone or insulative residues
No special handling requirements for shipping and storage; may be used in normal workplace environments. Cleaners with silicone leave an insulative layer preventing conductive or dissipative mats from functioning properly.
Certifications: Lead-free RoHS compliant.