Order Entry
Switzerland
ContactUsLinkComponent
Lid for 10 L Bucket
Lid for 10 L Bucket
Catalog # USBE4510
Lid for 10 L Bucket
Catalog # USBE4510

Specifications

  • Description:
    Lid for 10 L bucket, stainless steel
  • Pk:
    1
  • Ø:
    295 mm

Specifications

About this item

Stainless steel.