4-Vinyl-1,3-dioxolan-2-one
Supplier: Thermo Fisher Scientific
Jump to:
About this item
- CAS Number:4427-96-7
- Formula:C₅H₆O₃
- MDL Number:MFCD00143315
- Molecular Weight:114.10 g/mol
- Boiling Pt:237 °C
- Density:1.188
- Flash Pt:96 °C (205 °F)
- Flash Pt:96 °C(205 °F)
- Storage Temperature:Ambient
- UN:2810
- ADR:6.1,III
- ADR:"6.1",III
2 Options Available Below
Product Details & Documents
- CAS Number:4427-96-7
- Formula:C₅H₆O₃
- MDL Number:MFCD00143315
- Molecular Weight:114.10 g/mol
- Boiling Pt:237 °C
- Density:1.188
- Flash Pt:96 °C (205 °F)
- Flash Pt:96 °C(205 °F)
- Storage Temperature:Ambient
- UN:2810
- ADR:6.1,III
- ADR:"6.1",III



