Stands for Transferpette® electronic Pipettes
Anbieter: Brand
Zertifikate
Über diesen Artikel
Save space in the laboratory and use stands to safely store your BRAND single or multichannel pipettes.
The single stand is ideal for storing one Transferpette® electronic pipette - one stand for pipettes up to 1000 µl and another for the 5 mL pipette.
Three Transferpette® electronic pipettes (single channel, up to 1000 μl) can be charged simultaneously in the charging stand. A separate power supply is no longer needed.
Frequently Bought Together