Order Entry
Switzerland
ContactUsLinkComponent
High temperature tube furnaces with Tmax 1600 °C, RHTC series
High temperature tube furnaces with Tmax 1600 °C, RHTC series
Supplier:  NABERTHERM
Certificates
High temperature tube furnaces with Tmax 1600 °C, RHTC series
Supplier:  NABERTHERM

Specifications

  • Catalog No:
  • 455-2509
  • 455-2510
  • 455-2511
  • Tube ext.Ø:
  • 80 mm
  • 80 mm
  • 80 mm
  • Weight:
  • 50 kg
  • 70 kg
  • 90 kg

Specifications

Frequently Bought Together