ContactUsLinkComponent
Accessories for laboratory jacks
Accessories for laboratory jacks
Catalog # 245-1225
Accessories for laboratory jacks
Catalog # 245-1225
Specifications
  • For:
    245-1221-245-1228
  • With:
    M 10 thread
  • Description:
    Clamp, three-pronged brass, nickel-plated with vinyl-coated jaws and flexible shaft
  • Pk:
    1