Order Entry
Switzerland
ContactUsLinkComponent
2-Chloro-6-fluorobenzoic acid ≥98.0% for synthesis, Sigma-Aldrich®
Irritant
2-Chloro-6-fluorobenzoic acid ≥98.0% for synthesis, Sigma-Aldrich®
Catalog # 8.41200.0025
Supplier:  Merck
CAS Number:  
Irritant
2-Chloro-6-fluorobenzoic acid ≥98.0% for synthesis, Sigma-Aldrich®
Catalog # 8.41200.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay (acidimetric):
    ≥ 98.0 %
  • Melting range (lower value):
    ≥ 156 °C
  • Melting range (upper value):
    ≤ 159 °C
  • Identity (IR):
    passes test

Specification Test Results

About this item

 :
 :174.56 g/mol
Boiling Pt:272 °C (1013 hPa)
Melting Pt:157…159 °C
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:207-105-2
 :GHS07
CAS Number:434-75-3
Formula:C₇H₄ClFO₂
MDL Number:MFCD00002417