Order Entry
Switzerland
ContactUsLinkComponent
 
Microplate readers, Infinite® 200 PRO
 :  Tecan
 
undefined
Microplate readers, Infinite® 200 PRO
 :  Tecan

 

  • Catalog No:
  • TCAT92100004
  • TCAT92000004
  • TCAT92500004
  • TCAT92400004
  • TCAT92300004
  • TCAT92200004
  • Model:
  • Infinite® M Nano+
  • Infinite® M Plex
  • Infinite® F Plex
  • Infinite® F Nano+
  • Infinite® Lumi
  • Infinite® M Nano
  • Measuring modes:
  • Absorbance, Fluorescence
  • Absorbance, Fluorescence, Luminescence (glow, flash, dual-color)
  • Absorbance, Fluorescence, Luminescence (glow, falsh, dual-color), TR-FRET, HTRF® and FP
  • Absorbance, Fluorescence
  • Luminescence (glow, flash, dual-color)
  • Absorbance
  • Wavelength range:
  • Absorbance: 230 - 1000 nm
    FI Excitation: 230 - 850 nm
    FI Emission: 280 - 850 nm
  • Absorbance: 230 - 1000 nm
    FI Excitation: 230 - 850 nm
    FI Emission: 280 - 850 nm
    Luminescence: 380 - 600 nm
  • Absorbance: 230 - 1000 nm,
    FI Excitation: 230 - 850 nm,
    FI Emission: 280 - 850 nm,
    Luminescence: 380 - 600 nm
  • Absorbance: 230 - 1000 nm,
    FI Excitation: 230 - 850 nm,
    FI Emission: 280 - 850 nm
  • Luminescence: 380 - 600 nm
  • Absorbance: 230 - 1000 nm
  • Wavelength selection:
  • Monochromator optics
  • Monochromator optics
  • Filter optics
  • Filter optics
  • Filter-optics for Luminescence
  • Monochromator optics
  • Languages:
  • English, German, French, Italian, Spanish, Portuguese, Czech, Russian
  • English, German, French, Italian, Spanish, Portuguese, Czech, Russian
  • English, German, French, Italian, Spanish, Portuguese, Czech, Russian
  • English, German, French, Italian, Spanish, Portuguese, Czech, Russian
  • English, German, French, Italian, Spanish, Portuguese, Czech, Russian
  • English, German, French, Italian, Spanish, Portuguese, Czech, Russian
  • Plate types:
  • 6 - 384 well plates
  • 6 - 384 well plates
  • 6 - 384 well plates
  • 6 - 384 well plates
  • 6 - 384 well plates
  • 6 - 384 well plates
  • Shaking:
  • Linear and orbital shaking
  • Linear and orbital shaking
  • Linear and orbital shaking
  • Linear and orbital shaking
  • Linear and orbital shaking
  • Linear and orbital shaking
  • Temperature control:
  • 5 °C above ambient to 42 °C
  • 5 °C above ambient to 42 °C
  • 5 °C above ambient to 42 °C
  • 5 °C above ambient to 42 °C
  • 5 °C above ambient to 42 °C
  • 5 °C above ambient to 42 °C

 

Frequently Bought Together