Union Fittings, Hose Barb, Tee, Polypropylene, Dynalon®
Anbieter: Avantor Fluid Handling
Zertifikate
Über diesen Artikel
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Environmentally Preferable:
Technische Daten
- Orifice dimensions:
- 10 mm
- 5 mm
- 7 mm
- 2 mm
- Sterilisation:
- Autoclave only
- Autoclave only
- Autoclave only
- Autoclave only
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
Technische Daten
Frequently Bought Together