Order Entry
Switzerland
ContactUsLinkComponent
Staphylococcus aureus NCTC 6571 Lenticule® discs, 30-120 CFU, for microbiology, certified reference material
Staphylococcus aureus NCTC 6571 Lenticule® discs, 30-120 CFU, for microbiology, certified reference material
Catalog # CRM06571L-10EA
Supplier:  Merck
CAS Number:  
Staphylococcus aureus NCTC 6571 Lenticule® discs, 30-120 CFU, for microbiology, certified reference material
Catalog # CRM06571L-10EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Description:
    Staphylococcus Aureus Discs, 30-120 CFU NCTC 6571 Lenticule®, CRM
  • M-Clarity quality level:
    MQ300
  • Storage temperature:
    –20 °C
  • Product type:
    Hydrosoluble discs

Specifications

About this item

Certified Reference Microorganisms

  • Ready-to-use microbiological controls minimise the need for maintaining control strains in the test laboratory
  • Defined CFU range and low standard deviation
  • Fast, reliable and easy to use
  • From cultures provided by recognised Biological Resource Centres (BRCs) such as NCTC® and CECT®
  • Shelf life of 1-3 years

Vitroids™ and LENTICULE® discs contain viable microorganisms in a certified quantity (generally accredited according ISO/IEC 17025), produced under reproducible conditions compliant with ISO 17034:2016 using authenticated strains from NCTC®, NCPF® and CECT®.
Consisting of pure cultures of bacteria, fungi or yeasts in a solid water soluble matrix, they are stable for at least one year and are in a viable state with a shelf life of 1-3 years. The within-batch variation for every product is very low. Each batch is provided with a comprehensive certificate of analysis that specifies the mean number of colony forming units (CFU), an expanded uncertainty about the mean, details about the method used to determine the product data and the number of passages (subcultures) from the original strain.

Certifications: Manufactured under conditions compliant with ISO 2016:17034