

Dichloroisocyanuric acid sodium salt dihydrate, AnalaR NORMAPUR® analytical reagent
Catalog # 86197.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay (iodometric) (calculated on dried):≥ 98.0 %
- Heavy metals (as Pb):≤ 10 ppm
- Loss on drying:9.0 - 16.0 %
- Fe (Iron):≤ 50 ppm
Specification Test Results
About this item
:
:255.98 g/mol
Storage Temperature:Ambient
EINECS:220-767-7
UN:3077
ADR:9,
:GHS07
:GHS09
CAS Number:51580-86-0
Formula:C₃Cl₂N₃NaO₃·2H₂O
MDL Number:MFCD00149404