Auftragserfassung
Switzerland
ContactUsLinkComponent
VWR®, GF AAS Multi element standard XVIII, 16 components in 5% HNO₃
VWR®, GF AAS Multi element standard XVIII, 16 components in 5% HNO₃
Catalog # 91265.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, GF AAS Multi element standard XVIII, 16 components in 5% HNO₃
Catalog # 91265.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Packung:
    HDPE bottle
  • VE:
    100 mL
  • Bezeichnung:
    GF AAS Multi element standard XVIII (16 components)
  • Seq:
    02
  • Beschreibung Gefahrenklasse:
    Corrosive
  • UN No.:
    3264
  • Lagertemperatur:
    Ambient
  • Elements and Concentration:
    Ag 10 mg/l, Al 100 mg/l, As 100 mg/l, Ba 50 mg/l, Be 5 mg/l, Cd 5 mg/l, Co 50 mg/l, Cr 20 mg/l, Cu 50 mg/l, Fe 20 mg/l, Mn 20 mg/l, Ni 50 mg/l, Pb 100 mg/l, Sb 100 mg/l, Se 100 mg/l, Tl 100 mg/l

Specifications

About this item

16 components: Ag 10 mg/l, Al 100 mg/l, As 100 mg/l, Ba 50 mg/l, Be 5 mg/l, Cd 5 mg/l, Co 50 mg/l, Cr 20 mg/l, Cu 50 mg/l, Fe 20 mg/l, Mn 20 mg/l, Ni 50 mg/l, Pb 100 mg/l, Sb 100 mg/l, Se 100 mg/l, Tl 100 mg/l in 5% HNO₃

Certifications: ISO 17025/17034