Metal-in static shielding bags, SCS
: DESCO
Transparent metal-in static shielding bag provides a static safe environment for ESD sensitive electronic devices (ESDS).
- Printed with an ESD protective symbol and a lot code for traceability
- Heat sealable or zip-locked
- Manufactured from a polyester, metal, polyethylene laminate
The polyester dielectric in concert with the metal layer provides discharge shielding. The exterior being static dissipative allows electrostatic charges to be removed when grounded.
: Complies with IEC 61340-5-1 and IEC 61340-5-3.
: Suppied per pack of 100 bags in an oversized bag. Case quantities available in 'pizza box' style corrugated cardboard.
Frequently Bought Together