ContactUsLinkComponent
Face shields, V5 series
Face shields, V5 series
Catalog # MMMA5A-CP
Supplier:  3M
undefined
Face shields, V5 series
Catalog # MMMA5A-CP
Supplier:  3M
Specifications
  • Colour:
    Transparent
  • Description:
    5A-CP face shield with acetate visor
  • Pk:
    1
Specifications
About this item
Acetate face shields are lightweight and non-conductive.
  • Acetate visor
  • Lightweight
  • Non-conductive
3M™ headgear and face shields help provide head and eye protection against heat, debris, and chemicals. The 5A-CP offers chemical and impact resistance with its acetate face shield and its chin cup provides extended protection with downward visibility.