VWR®, Withdrawal System for Solvents with Manual Pressure Build-up for 10 and 30 L Stainless Steel Barrels
99900.001
- Description:Withdrawal system for solvents with manual pressure build-up for 10 and 30 L stainless steel barrels
- Environmentally Preferable:
- Pk:1
Specially developed solvent withdrawal system with manual pressure build-up for VWR® returnable 10 and 30 L stainless steel barrels.
- Two dip tubes
- Hand pump ball
- Perfluorelastomer o-rings
This set contains a specially developed solvent withdrawal system for manual pressure build-up which can be used with VWR® 10 and 30 L returnable stainless steel barrels. This set includes two dip tubes, one for 10 L and one for 30 L barrels as well as a hand pump ball with a brass coupling.
: Complete set contains withdrawal system for 10 and 30 L barrels (stainless steel), Perfluorelastomer O-rings, 2 dip tubes (one for 10 L and one for 30 L), and hand pump ball with brass coupling.