ContactUsLinkComponent
Knife storage wall cabinet
Knife storage wall cabinet
Codice catalogo TEKMSHCB-WCSK45X20
Nome del fornitore:  TEKNOMEK
Knife storage wall cabinet
Codice catalogo TEKMSHCB-WCSK45X20
Nome del fornitore:  TEKNOMEK
Specifiche
  • Description:
    Knife storage wall cabinet
  • Material:
    Stainless steel 304
  • W×D×H:
    450×200×600 mm
  • Pk:
    1
Specifiche
Informazioni su questo articolo
High quality lockable storage cabinet for knives.
  • Includes a replaceable magnetic knife holder
  • Brushed dull polish finish
  • Wall fixed
  • Lockable
  • Sloping top
Our knife storage cabinet is wall mounted and manufactured from 304 grade stainless steel with a brushed dull polish finish. With a lockable door and a replaceable magnetic block for secure storage of knives, this cabinet is perfect for secure and hygienic utensil storage.
The sloping top is extremely hygienic and is very easy to clean.