Chemical resistant gloves, AlphaTec® 87-108
: Ansell
Long-cuff natural latex rubber gloves, offering heavy-duty chemical and mechanical protection.
- Tear-resistant external surface for improved chemical protection, particularly against water-based hazards
- Long beaded cuff enhances wearer safety and facilitates donning
- Maximum glove length of 610 mm/23” ensuring additional forearm and wrist protection
- Chlorination treatment makes these gloves harder and cleanses their surface
- Natural latex rubber formulation means these gloves maintain flexibility and elasticity, despite their thickness
Recommended for use in: Agriculture, construction, janitorial use, chemical processing and preparation, maintenance of plant and machines, sandblast cleaning.
: EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5, EN 388:2016 (3121B); Cat. III; AQL 0,65
- Catalog No:
- ANSE87-108/8.5
- ANSE87-108/7.5
- ANSE87-108/9.5
- ANSE87-108/10.5
- Beidseitig tragbar / handspezifisch:
- Handspezifisch
- Handspezifisch
- Handspezifisch
- Handspezifisch
- Einweg / wiederverwendbar:
- wieder verwendbar
- wieder verwendbar
- wieder verwendbar
- wieder verwendbar
- Latex / ohne Latex:
- Latex
- Latex
- Latex
- Latex
- Gepudert/ungepudert:
- Ungepudert
- Ungepudert
- Ungepudert
- Ungepudert
- Material:
- Naturkautschuk-Latex
- Naturkautschuk-Latex
- Naturkautschuk-Latex
- Naturkautschuk-Latex
- Oberfläche:
- Smooth
- Smooth
- Smooth
- Smooth
- Stulpenart:
- Rollrand
- Rollrand
- Rollrand
- Cuff_Style-21391117|Cuff_Style-21391103|Cuff_Style-21391099
- Stärke - Handfläche:
- 1,5 mm
- 1,5 mm
- 1,5 mm
- 1,5 mm
- EN Standard:
- EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5, EN 388:2016 (3121B)
- EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5, EN 388:2016 (3121B)
- EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5, EN 388:2016 (3121B)
- EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5, EN 388:2016 (3121B)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
Frequently Bought Together