Industrial gloves, EDGE® 48-700
Anbieter: Ansell
Zertifikate
Über diesen Artikel
These comfortable high-dexterity gloves are designed with a seamless liner and knitted wrist to ensure a good level of flexibility and close fit, and may also be worn as an underglove.
- Liner made from PE/wrapped yarn/fiber glass/polyester/Spandex
- Good level of dexterity and flexibility
- High level of cut and abrasion resistance
- May be used as a liner under another glove
- Reinforced thumb crotch for increased abrasion resistance and longer performance
- Compliant with FDA food handling requirements
Recommended for: Slicing, cutting and shopping; cleaning and changing slicer blades in delis and supermarkets; defeathering and skinning; assembly and finishing; body assembly; machine tool operation; sheet metals and metalwork.
Compliant with FDA food handling requirements. Compliant with the following provisions: the EC-regulation 1935/2004 and the EC-regulation 2023/2006 related to Good Manufacturing Practices (GMP) for Materials and Articles intended to come in contact with Foodstuffs.
Certifications: EN 388:2016 (3X4XC), Cat. II
Food contact approved.
Food contact approved.
Technische Daten
- Catalog No:
- ANSE48-700/9
- ANSE48-700/7
- ANSE48-700/8
- ANSE48-700/10
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Gauge:
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- FDA regulated:
- +
- +
- +
- +
- EN norm:
- EN 388:2016 (3X4XC)
- EN 388:2016 (3X4XC)
- EN 388:2016 (3X4XC)
- EN 388:2016 (3X4XC)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Food contact approved:
- +
- +
- +
- +
Technische Daten
Frequently Bought Together