About this item
Clean and Sterile disposable boot covers are individually double-bagged in pairs and folded to promote easier gowning with less excursions.
- Clean and sterile in silicone-free environment
- Latex-free
- Tunnelled elastic top
- 19" High
- Non-skid PVC sole
- Individually double-bagged in pairs
- Gamma radiation indicator dots on each package
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparel provides chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Specifications
- Catalog No:
- 114-9549
- 114-9548
- 114-9550
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom