ContactUsLinkComponent
MEA cleanroom looped face mask, BioClean™
MEA cleanroom looped face mask, BioClean™
Catalog # NITRMEA210-2
Supplier:  Nitritex
undefined
MEA cleanroom looped face mask, BioClean™
Catalog # NITRMEA210-2
Supplier:  Nitritex
Specifications
  • Type:
    With earloops
  • Colour:
    White
  • Environmentally Preferable:
  • Pk:
    600
Specifications
About this item
Non sterile ear loop face mask.
  • High bacterial, viral, and particle efficiency filtration
  • Fully enclosed malleable noseband
  • Ultrasonically sealed edges
  • Ear loops for quick and easy donning
  • Clip connector for secure fastening at back of head available to purchase separately
Providing high particle and bacterial filtration efficiency, the BioClean™ MEA Cleanroom Looped Face Mask is manufactured from cleanroom compatible materials to reduce contamination into the controlled environment. The mask features ear loops for quick donning and doffing. (Clip connector to allow for quick and secure fastening at back of head are available to purchase separately; see datasheet).