Multi-purpose medium-duty gloves, ActivArmr® 97-008
Anbieter: Ansell
Zertifikate
Über diesen Artikel
Medium-duty industrial gloves, offering durable hand coverage with assured grip.
- Comfortable and breathable
- Foam nitrile coating enhances grip performance in wet or dry conditions
- Seamless knit design and reinforced ridges for excellent tactile sensitivity without sacrificing comfort
- Adjustable wrist closure with pull-tab as well as an ID nameplate
- DuPont™ Kevlar® layer ensures tough, durable protection against cuts (ANSI A2/EN ISO B) and harsh abrasion (ANSI and EN level 4)
Suitable for a wide range of medium-duty tasks such as body assembly, general maintenance, engine transmission and repair as well as hydraulic repair.
Certifications: EN 388:2016 (4232B), Cat. II
Technische Daten
- Catalog No:
- ANSE97-008/9
- ANSE97-008/11
- ANSE97-008/8
- ANSE97-008/10
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Nylon/Spandex/Kevlar®
- Nylon/Spandex/Kevlar®
- Nylon/Spandex/Kevlar®
- Nylon/Spandex/Kevlar®
- Finish:
- Palm coated (foam nitrile)
- Palm coated (foam nitrile)
- Palm coated (foam nitrile)
- Palm coated (foam nitrile)
- Gauge:
- 15
- 15
- 15
- 15
- Cuff style:
- Hook & loop/Velcro® fastening
- Hook & loop/Velcro® fastening
- Hook & loop/Velcro® fastening
- Hook & loop/Velcro® fastening
- EN norm:
- EN 388:2016 (4232B)
- EN 388:2016 (4232B)
- EN 388:2016 (4232B)
- EN 388:2016 (4232B)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
Technische Daten
Frequently Bought Together