ContactUsLinkComponent
Industrial gloves, HyFlex® 11-422
Industrial gloves, HyFlex® 11-422
Supplier:  Ansell
Certificates
undefined
Industrial gloves, HyFlex® 11-422
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE11-422/10
  • ANSE11-422/7
  • ANSE11-422/9
  • ANSE11-422/8
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex
  • Latex
  • Latex
  • Latex
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Palm coating (PU/nitrile)
  • Palm coating (PU/nitrile)
  • Palm coating (PU/nitrile)
  • Palm coating (PU/nitrile)
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X32B)
  • EN 388:2016 (4X32B)
  • EN 388:2016 (4X32B)
  • EN 388:2016 (4X32B)
  • Food contact approved:
  • +
  • +
  • +
  • +