ContactUsLinkComponent
Cleanroom hoods, CleanMax®
Cleanroom hoods, CleanMax®
Catalog # 114-9543
undefined
Cleanroom hoods, CleanMax®
Catalog # 114-9543
Specifications
  • Clothing Size:
    One size
  • Garment application:
    Cleanroom
  • Colour:
    White
  • Seq:
    01
  • Pk:
    100 PAIR
  • Cleanroom class:
    100/ISO 5
Specifications
About this item
Clean manufactured disposable apparel are individually packaged and folded to promote easier gowning with less excursions.
  • Clean manufactured in silicone-free environment
  • Latex-free
  • Tunnelled elastic on wrists with thumb loops
  • Zipper closure
  • For critical environments
  • Individually bagged
CleanMax® cleanroom garments provide comfort, quality, and protection.
All apparel provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunnelled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certifications: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.
Packaging: 100/case, individually packed.