ContactUsLinkComponent
Cleanroom frocks/lab coats, CleanMax®
Zertifikate
undefined
Cleanroom frocks/lab coats, CleanMax®
Technische Daten
  • Catalog No:
  • 114-9563
  • 114-9568
  • 114-9567
  • 114-9569
  • 114-9566
  • 114-9565
  • 114-9564
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Cleanroom class:
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
Technische Daten
Frequently Bought Together