Accessories for Rotary Vacuum Concentrator, RVC 2-25 CDplus universal package
CHRS110369
- Description:Angular rotor made of aluminium, black anodized, tube dimensions Ø 12×36 mm with flat base, setting angle 40°
- Pk:1
- For:Total capacity 3 rotors for a total of 108 vessels