



Benzyltrimethylammonium hydroxide 40% in methanol for synthesis, Sigma-Aldrich®
8.01816.5000
: Merck
:
- Pack type:PE/metal barrel
- Pk:5 L
- Seq:5190
- Solubility:(20 °C) soluble
- Melting Point:-98 °C
- Boiling Point:65 °C (1013 hPa)
- Vapor Pressure:127 hPa (20 °C)
- Flash Point:15 °C
- Explosion Limit:5.1 - 44 %(V)
- Density:0.93 g/cm3 (20 °C)
- pH:12 (H2O, 20 °C) (undiluted)
- Ignition Point:455 °C
- Synonyms:Trimethylbenzylammonium hydroxide, Triton B
:
:167.25
:3 (6.1, 8),II
:M-Clarity™ quality level = MQ200
:3286
:GHS02
:GHS06
:GHS05
:GHS08
:100-85-6
:C₁₀H₁₇NO
:MFCD00008281