Order Entry
Switzerland
ContactUsLinkComponent
Manganese(II) sulphate monohydrate ≥98.0%, powder BP, FCC
Health
Manganese(II) sulphate monohydrate ≥98.0%, powder BP, FCC
Catalog # SPCMMA164-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Health
Manganese(II) sulphate monohydrate ≥98.0%, powder BP, FCC
Catalog # SPCMMA164-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (MnSO₄[-H₂O):
    98.0 - 102.0%
  • Assay (Ignited):
    99.0 - 101.0%
  • Loss On Heating:
    10.0 - 12.0%
  • Appearance of Solution:
    To Pass Test
  • Arsenic (As):
    max. 3 mg/kg

Specification Test Results

About this item

 :
ADR:9,III
 :169.02 g/mol
UN:3077
 :GHS08
Storage Temperature:Ambient
CAS Number:10034-96-5
Formula:MnSO₄·1H₂O
MDL Number:MFCD00149159