

Copper(II) sulphate pentahydrate ≥98.0%, granular FCC
:7758-99-8
:
:CuSO₄·5H₂O
:MFCD00149681
:249.69 g/mol
:3077
:9,III
:GHS06
:GHS07
:Ambient
- Assay (CuSO₄.5H₂O):98.0 - 102.0 %
- Iron (Fe):max. 0.01 %
- Lead (Pb):max. 4 mg/kg
- Substances Not Pptd.By Hydrogen Sulfide:max. 0.3 %
- Identification:To Pass Test
Frequently Bought Together