Order Entry
Switzerland
ContactUsLinkComponent
2,2'-Methylenebis(6-tert-butyl-p-cresol) for synthesis, Sigma-Aldrich®
Health
2,2'-Methylenebis(6-tert-butyl-p-cresol) for synthesis, Sigma-Aldrich®
Catalog # 8.18447.9045
Supplier:  Merck
CAS Number:  
Health
2,2'-Methylenebis(6-tert-butyl-p-cresol) for synthesis, Sigma-Aldrich®
Catalog # 8.18447.9045
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    45 kg
  • Seq:
    4388

Specifications

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    125 - 128 °C
  • Molar Mass:
    340.5 g/mol
  • Bulk Density:
    300 kg/m3
  • Boiling Point:
    187 °C (0.7 hPa)
  • Flash Point:
    195 °C
  • Density:
    1.07 g/cm3 (20 °C)
  • pH:
    4 - 7 (H2O, 20 °C) (aqueous suspension)
  • Ignition Point:
    350 °C
  • Synonyms:
    2,2'-Methylenebis-(6-tert-butyl-4-methyl-phenol), Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)-methan
  • Hill Formula:
    C23H32O2
  • Chemical Formula:
    (2-(OH)-3-(CH3)3C-5-(CH3)C6H2)2CH2

Specification Test Results

About this item

 :
 :340.51 g/mol
 :M-Clarity™ quality level = MQ200
 :GHS08
CAS Number:119-47-1
Formula:C₂₃H₃₂O₂
MDL Number:MFCD00043641