Order Entry
Switzerland
ContactUsLinkComponent
Hydroxypropyl methylcellulose (Hypromellose), 4000 mPa.s USP Substitution Type 2910
Hydroxypropyl methylcellulose (Hypromellose), 4000 mPa.s USP Substitution Type 2910
Catalog # SPCMH1335-125GM
Supplier:  Spectrum Chemical
CAS Number:  
Hydroxypropyl methylcellulose (Hypromellose), 4000 mPa.s USP Substitution Type 2910
Catalog # SPCMH1335-125GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    125 g
  • Seq:
    3445
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Viscosity of a 2% Aqueous Solution @ 20D °C:
    3000- 5600 mPa·s
  • Assay (Methoxy Groups):
    28.0 - 30.0%
  • Assay (Hydroxypropoxy Groups):
    7.0 - 12.0%
  • pH:
    5.0 - 8.0
  • Loss On Drying:
    max. 5.0%

Specification Test Results

About this item

Synonyms: Methocel K100, Hydroxypropyl methyl cellulose
 :
Storage Temperature:Ambient
CAS Number:9004-65-3
Formula:C₃H₈O₂·xCH₄O
MDL Number:MFCD00131360