Order Entry
Switzerland
ContactUsLinkComponent
Calcium sulphate dihydrate ≥98.0% FCC
Calcium sulphate dihydrate ≥98.0% FCC
Supplier:  Spectrum Chemical
Certificates
Calcium sulphate dihydrate ≥98.0% FCC
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay (Dried Basis):
    98.0 % min.
  • Loss On Drying:
    19.0 - 23.0 %
  • Fluoride:
    max. 0.003 %
  • Lead (Pb):
    max. 2 mg/kg
  • Selenium (Se):
    max. 0.003 %

Specification Test Results

Frequently Bought Together