Structural adhesive nozzles, Scotch-Weld™
Catalog # MMMAD400
Supplier: 3M
Specifications
- Description:Structural adhesive nozzles
- Pk:36
Specifications
About this item
3M™ Scotch-Weld™ Structural adhesive nozzles are for use with 3M EPX™ Duo-Pak adhesives to ensure correct mixing of adhesive parts. Nozzle is matched to adhesive.
- Get high performance adhesive matching
- Use with 3M EPX™ Duo-Pak adhesives
- Apply to low volume applications
- Easy and accurate application
- Can be replaced by new nozzles for future applications
Static mixing nozzles for 3M™ EPX™ Adhesives. Mixing nozzles allow for precise mixing of the adhesive and activator and easy and accurate application. Mixing nozzles can be used throughout the work life of the adhesive mix and can be disposed of and replaced by new nozzles for future applications.