ContactUsLinkComponent
2,6-Di-tert-butyl-p-cresol ≥99.0%, granular FCC
Irritant
2,6-Di-tert-butyl-p-cresol ≥99.0%, granular FCC
Supplier:  Spectrum Chemical
Certificates
Irritant
2,6-Di-tert-butyl-p-cresol ≥99.0%, granular FCC
Supplier:  Spectrum Chemical
Specification Test Results
  • Assay (C₁₅H₂₄O):
    99.0 % min.
  • Residue On Ignition:
    max.0.002 %
  • Lead:
    max. 2 mg/kg
  • Identification:
    To Pass Test