ContactUsLinkComponent
Cleanroom bouffant caps, Tyvek® IsoClean®, Model IC729B WS
Cleanroom bouffant caps, Tyvek® IsoClean®, Model IC729B WS
Katalognr. DUPOIC729SWHWS
Supplier:  DuPont
undefined
Cleanroom bouffant caps, Tyvek® IsoClean®, Model IC729B WS
Katalognr. DUPOIC729SWHWS
Supplier:  DuPont
Specifications
  • Sterile:
    +
  • Colour:
    White
  • Pk:
    250
Specifications
About this item
These bouffant caps, model IC729SWH WS, are made of Tyvek®, which is a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability, and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
  • Ultra-low linting fabric
  • Serged seams
  • Elastic headband
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
  • Suitable for use in GMP class A/B (ISO Class 5) cleanrooms
Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
All DuPont™ Tyvek® IsoClean® sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy-tear cleanroom bag.
Garments and accessories made of sterile Tyvek® IsoClean® are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry, as well as in other critical or controlled environments.
Certifications: PPE Category I.
Packaging: 50 per box, individually packed. Subgrouped by 50 in an outer bag. Two polyethylene liners. Cardboard box.