ContactUsLinkComponent
Hardness (Ca and total sequential) HR reagent set, digital titrator
Hardness (Ca and total sequential) HR reagent set, digital titrator
Catalog # HACH2448700
Supplier:  Hach
CAS Number:  
undefined
Hardness (Ca and total sequential) HR reagent set, digital titrator
Catalog # HACH2448700
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    EDTA
  • Pk:
    1 KIT
  • Description:
    No of tests: 100
  • Range:
    100 - 4000 mg/L CaCO₃
Specifications
About this item
For sequential determination of calcium and total hardness by EDTA digital titrator titration.
  • Accurate results; when and where you need them
  • Fast and flexible
  • Versatile
  • Economical
Hach method 8329. Range: 100 to 4000 mg/L CaCO₃. Set contains buffer solution (Hardness 1), CalVer® 2 calcium indicator powder pillows, ManVer® 2 hardness indicator powder pillows, potassium hydroxide solution (8 N), sulfuric acid (5,25 N) and 0,800 M EDTA digital titrator cartridge. Approximately 40 to 100 tests.
Ordering information: Hazardous items; items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.