Toxtrak™ toxicity reagent set
Katalognr. HACH2597200
Anbieter: Hach
CAS-Nummer:
Technische Daten
- Pk:1 SET
- Description:Toxtrak™ toxicity reagent set
- Range:25 - 49
Technische Daten
Über diesen Artikel
Use the toxtrak™ toxicity reagent set to analyze your water or wastewater samples for toxicity.
- The toxtrak™ reagents perform toxicity screens on drinking water, wastewater and natural water
- Toxicity is compared to a baseline using indigenous biomass or stock bacteria cultures with a range of zero 100% inhibition
- A colorimetric method is used to determine the toxicity of samples, with analysis done using one of the hach's trusted spectrophotometers
Follows hach toxicity method 10017.
Ordering information: Set includes total bacteria count media set (15 TBC tubes, 15 individually wrapped sterile pipets), toxtrak™ powder pillows (pk/50), toxtrak™ accelerator solution (15 ml), disposable culture tubes (5 packs; each pk/6), sterile transfer pipets (pk/15) and sodium thiosulfate standard (100 ml, 0,0246N). Zero to 100% inhibition.